siRNA Details
| Gene Symbol: Ankrd45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028664.1 | NP_082940.1 | 2 | 301 | ||
| GenBank mRNA | BC049713.1 | - | 2 | 273 | 6741553 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK144074.1 | - | 2018 | Chr. - - [-] on Build | ||

Ankrd45
Genomic Map