siRNA Details
| Gene Symbol: 4930415O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201322.1 | NP_001188251.1 | 2 | 224 | ||
| GenBank mRNA | AK015155.1 | - | 2 | 208 | ||
| AK029626.1 | - | 2 | 208 | |||
| BC048564.1 | - | 2 | 224 | 6705104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930415O20Rik| Gene Symbol: 4930415O20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201322.1 | NP_001188251.1 | 2 | 224 | ||
| GenBank mRNA | AK015155.1 | - | 2 | 208 | ||
| AK029626.1 | - | 2 | 208 | |||
| BC048564.1 | - | 2 | 224 | 6705104 | ||