siRNA Details
| Gene Symbol: Zdbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267872.1 | NP_001254801.1 | 6 | 4744 | ||
| NM_001285936.1 | NP_001272865.1 | 7 | 4878 | |||
| NM_001285937.1 | NP_001272866.1 | 6 | 4821 | |||
| NM_028673.1 | NP_082949.1 | 7 | 4794 | |||
| GenBank mRNA | AB777270.1 | - | 7 | 4878 | ||

Zdbf2
Genomic Map