siRNA Details
| Gene Symbol: Fam151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163627.1 | NP_001157099.1 | 5, 6 | 758 | ||
| GenBank mRNA | AK015100.1 | - | 5, 6 | 741 | ||
| AK162422.1 | - | 4, 5 | 577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam151b| Gene Symbol: Fam151b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163627.1 | NP_001157099.1 | 5, 6 | 758 | ||
| GenBank mRNA | AK015100.1 | - | 5, 6 | 741 | ||
| AK162422.1 | - | 4, 5 | 577 | |||