siRNA Details
| Gene Symbol: Als2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159948.2 | NP_001153420.2 | 12 | 2656 | ||
| NM_028717.6 | NP_082993.4 | 12 | 2487 | |||
| NM_146109.3 | NP_666221.1 | 12 | 2487 | |||
| GenBank mRNA | AB053307.1 | - | 12 | 2475 | ||
| AK029858.1 | - | 12 | 2659 | |||
| BC031479.1 | - | 12 | 2464 | 3257574 | ||
| BC046828.1 | - | 12 | 2448 | 6409924 | ||
| CD543795.1 | - | 2 | 471 | |||
| CK634368.1 | - | 2 | 210 | |||

Als2
Genomic Map