siRNA Details
| Gene Symbol: Pop7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028753.2 | NP_083029.1 | 2 | 306 | ||
| GenBank mRNA | AK002782.1 | - | 2 | 308 | ||
| AK158620.1 | - | 1 | 218 | |||
| AK162268.1 | - | 2 | 311 | |||
| BC010780.1 | - | 2 | 220 | 4216163 | ||

Pop7
Genomic Map