siRNA Details
Gene Symbol: Slc35a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028756.4 | NP_083032.2 | 5, 6 | 777 | ||
GenBank mRNA | AK003124.1 | - | 5, 6 | 705 | ||
AK030566.1 | - | 5, 6 | 702 | |||
AK033921.1 | - | 5, 6 | 744 | |||
AK085567.1 | - | 5, 6 | 716 | |||
AK150825.1 | - | 5, 6 | 768 | |||
AK151484.1 | - | 5, 6 | 768 | |||
AK170026.1 | - | 5, 6 | 744 | |||
BC011115.1 | - | 5, 6 | 706 | 4013987 |
