siRNA Details
| Gene Symbol: Armc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166138.1 | NP_001159610.1 | 10 | 1075 | ||
| NM_028768.3 | NP_083044.2 | 10 | 1075 | |||
| GenBank mRNA | AK004793.2 | - | 10 | 1076 | ||
| AK078280.1 | - | 10 | 826 | |||
| AK083094.1 | - | 9 | 767 | |||
| BC038866.1 | - | 10 | 1033 | 1230321 | ||

Armc8
Genomic Map