siRNA Details
Gene Symbol: F13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166391.1 | NP_001159863.1 | 10, 11 | 1379 | ||
NM_028784.3 | NP_083060.2 | 10, 11 | 1404 | |||
GenBank mRNA | AK030914.1 | - | 11, 12 | 1379 | ||
AK036403.1 | - | 10, 11 | 1405 | |||
AK049092.1 | - | 10, 11 | 1405 | |||
AK165311.1 | - | 10, 11 | 1405 | |||
BC011073.1 | - | 4, 5 | 485 | |||
BC040274.1 | - | 10, 11 | 1395 | 3482233 |
