siRNA Details
| Gene Symbol: F13a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166391.1 | NP_001159863.1 | 6 | 777 | ||
| NM_028784.3 | NP_083060.2 | 6 | 802 | |||
| GenBank mRNA | AK030914.1 | - | 6 | 777 | ||
| AK036403.1 | - | 6 | 803 | |||
| AK049092.1 | - | 6 | 803 | |||
| AK163591.1 | - | 6 | 777 | |||
| AK165311.1 | - | 6 | 803 | |||
| AK170278.1 | - | 6 | 803 | |||
| BC040274.1 | - | 6 | 793 | 3482233 | ||

F13a1
Genomic Map