siRNA Details
| Gene Symbol: Prl8a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023741.2 | NP_076230.1 | 4 | 390 | ||
| GenBank mRNA | AF230921.1 | - | 4 | 376 | ||
| AF230922.1 | - | 4 | 379 | |||
| AF230923.1 | - | 5 | 592 | |||
| AF466150.1 | - | 4 | 391 | |||
| AK005568.1 | - | 4 | 396 | |||
| AK167369.1 | - | 4 | 397 | |||
| BC100326.1 | - | 4 | 384 | 30787396 | ||

Prl8a8
Genomic Map