siRNA Details
| Gene Symbol: Sipa1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081028.1 | NP_001074497.1 | 12 | 3764 | ||
| GenBank mRNA | AK163313.1 | - | 9 | 2146 | ||
| AK172968.1 | - | 6 | 645 | |||
| BC095969.1 | - | 2 | 297 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sipa1l3| Gene Symbol: Sipa1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081028.1 | NP_001074497.1 | 12 | 3764 | ||
| GenBank mRNA | AK163313.1 | - | 9 | 2146 | ||
| AK172968.1 | - | 6 | 645 | |||
| BC095969.1 | - | 2 | 297 | |||