siRNA Details
| Gene Symbol: Prss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193631.1 | NP_001180560.1 | 3 | 489 | ||
| GenBank mRNA | AK005740.1 | - | 3 | 494 | ||
| BC048657.1 | - | 3 | 489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss51| Gene Symbol: Prss51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193631.1 | NP_001180560.1 | 3 | 489 | ||
| GenBank mRNA | AK005740.1 | - | 3 | 494 | ||
| BC048657.1 | - | 3 | 489 | |||