siRNA Details
| Gene Symbol: Tex37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028825.3 | NP_083101.1 | 4 | 411 | ||
| GenBank mRNA | AK005862.1 | - | 4 | 412 | ||
| BC049736.1 | - | 4 | 389 | 6771350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex37| Gene Symbol: Tex37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028825.3 | NP_083101.1 | 4 | 411 | ||
| GenBank mRNA | AK005862.1 | - | 4 | 412 | ||
| BC049736.1 | - | 4 | 389 | 6771350 | ||