siRNA Details
| Gene Symbol: 1700016K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198637.2 | NP_941039.1 | 2 | 614 | ||
| GenBank mRNA | AK132806.1 | - | 2 | 539 | ||
| BC049758.1 | - | 2 | 553 | 6773542 | ||
| BQ931096.1 | - | 2 | 630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700016K19Rik| Gene Symbol: 1700016K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198637.2 | NP_941039.1 | 2 | 614 | ||
| GenBank mRNA | AK132806.1 | - | 2 | 539 | ||
| BC049758.1 | - | 2 | 553 | 6773542 | ||
| BQ931096.1 | - | 2 | 630 | |||