siRNA Details
Gene Symbol: Tubgcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286007.1 | NP_001272936.1 | 7 | 1202 | ||
NM_001286009.1 | NP_001272938.1 | 7 | 1202 | |||
NM_001286011.1 | NP_001272940.1 | 6 | 969 | |||
NM_133755.2 | NP_598516.2 | 7 | 1129 | |||
GenBank mRNA | AK033157.1 | - | 7 | 919 | ||
AK045084.1 | - | 7 | 918 | |||
AK146771.1 | - | 7 | 1124 | |||
AK153511.1 | - | 7 | 1050 | |||
AK158060.1 | - | 7 | 918 | |||
AK167924.1 | - | 7 | 1024 | |||
AK193646.1 | - | 2 | 38 | |||
AK194085.1 | - | 2 | 38 | |||
BC012519.1 | - | 7 | 1057 | 3601589 | ||
BC025564.1 | - | 6 | 891 | |||
BC025582.1 | - | 7 | 1018 | 5324981 | ||
BC027034.1 | - | 7 | 1113 |
