siRNA Details
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028833.3 | NP_083109.2 | 11 | 957 | ||
| GenBank mRNA | AK006472.2 | - | 10 | 698 | ||
| AK029333.1 | - | 10 | 721 | |||
| BC059223.1 | - | 11 | 957 | 6417270 | ||
| BC065128.1 | - | 9 | 638 | |||

Iqce
Genomic Map