siRNA Details
| Gene Symbol: Smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021535.4 | NP_067510.3 | 2 | 225 | ||
| GenBank mRNA | AB044414.1 | - | 2 | 154 | ||
| AK003493.1 | - | 2 | 212 | |||
| AK011140.1 | - | 2 | 224 | |||
| AK019174.1 | - | 2 | 184 | |||
| AK159635.1 | - | 2 | 212 | |||
| AK168813.1 | - | 2 | 219 | |||
| BC020034.1 | - | 2 | 167 | |||
| BC057446.1 | - | 2 | 186 | 3155760 | ||

Smu1
Genomic Map