siRNA Details
| Gene Symbol: 1700086L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007020.1 | - | 2 | 211 | ||
| BG083361.2 | - | 2 | 202 | |||
| BQ175117.1 | - | 3 | 618 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700086L19Rik| Gene Symbol: 1700086L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007020.1 | - | 2 | 211 | ||
| BG083361.2 | - | 2 | 202 | |||
| BQ175117.1 | - | 3 | 618 | |||