siRNA Details
Gene Symbol: 1700086L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005679.1 | - | 2 | 213 | ||
AK007020.1 | - | 3 | 314 | |||
BG083361.2 | - | 4 | 527 | |||
BQ175117.1 | - | 2 | 515 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700086L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005679.1 | - | 2 | 213 | ||
AK007020.1 | - | 3 | 314 | |||
BG083361.2 | - | 4 | 527 | |||
BQ175117.1 | - | 2 | 515 |