siRNA Details
| Gene Symbol: 1700092M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177347.1 | NP_001170818.1 | 1 | 75 | ||
| GenBank mRNA | AK007043.1 | - | 1 | 38 | ||
| BC048571.1 | - | 1 | 31 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700092M07Rik| Gene Symbol: 1700092M07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177347.1 | NP_001170818.1 | 1 | 75 | ||
| GenBank mRNA | AK007043.1 | - | 1 | 38 | ||
| BC048571.1 | - | 1 | 31 | |||