siRNA Details
| Gene Symbol: Mettl23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028865.3 | NP_083141.3 | 5 | 803 | ||
| GenBank mRNA | AK003450.1 | - | 5 | 649 | ||
| AK135294.1 | - | 930 | ||||
| AK160133.1 | - | 4 | 723 | |||
| AW490033.1 | - | 1 | 249 | |||
| BC026936.1 | - | 5 | 718 | 4021089 | ||

Mettl23
Genomic Map