siRNA Details
| Gene Symbol: Ranbp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145824.4 | NP_665823.2 | 2 | 492 | ||
| GenBank mRNA | AK048772.1 | - | 2 | 348 | ||
| AK173177.1 | - | 2 | 305 | |||
| AY337314.1 | - | 2 | 493 | |||
| BC024698.1 | - | 2 | 337 | 3667000 | ||
| EU281316.1 | - | 2 | 353 | |||

Ranbp10
Genomic Map