siRNA Details
| Gene Symbol: Smchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028887.3 | NP_083163.3 | 23 | 3077 | ||
| GenBank mRNA | AK129181.1 | - | 1 | 74 | ||
| BC062946.1 | - | 9 | 922 | 30357469 | ||
| BC076627.1 | - | 16 | 2040 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smchd1| Gene Symbol: Smchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028887.3 | NP_083163.3 | 23 | 3077 | ||
| GenBank mRNA | AK129181.1 | - | 1 | 74 | ||
| BC062946.1 | - | 9 | 922 | 30357469 | ||
| BC076627.1 | - | 16 | 2040 | |||