siRNA Details
| Gene Symbol: Spag17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028892.4 | NP_083168.3 | 3 | 369 | ||
| GenBank mRNA | AK016477.1 | - | 3 | 355 | ||
| AK029691.1 | - | 3 | 371 | |||
| AK029947.1 | - | 3 | 365 | |||
| AY555275.1 | - | 3 | 355 | |||
| AY792594.1 | - | 3 | 355 | |||

Spag17
Genomic Map