siRNA Details
| Gene Symbol: Spag17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028892.4 | NP_083168.3 | 36, 37 | 5489 | ||
| GenBank mRNA | AY792594.1 | - | 35, 36 | 5040 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spag17| Gene Symbol: Spag17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028892.4 | NP_083168.3 | 36, 37 | 5489 | ||
| GenBank mRNA | AY792594.1 | - | 35, 36 | 5040 | ||