siRNA Details
Gene Symbol: Spag17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028892.4 | NP_083168.3 | 46 | 6730 | ||
GenBank mRNA | AK136364.1 | - | 10 | 1234 | ||
AY792594.1 | - | 45 | 6281 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Spag17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028892.4 | NP_083168.3 | 46 | 6730 | ||
GenBank mRNA | AK136364.1 | - | 10 | 1234 | ||
AY792594.1 | - | 45 | 6281 |