siRNA Details
| Gene Symbol: Clec16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204229.1 | NP_001191158.1 | 3, 4 | 569 | ||
| NM_177562.5 | NP_808230.2 | 3, 4 | 569 | |||
| GenBank mRNA | AK089327.1 | - | 3, 4 | 500 | ||
| AK122255.1 | - | 3, 4 | 521 | |||
| AK155314.1 | - | 3, 4 | 511 | |||
| AK156927.1 | - | 3, 4 | 500 | |||
| AK170754.1 | - | 3, 4 | 500 | |||
| AK171134.1 | - | 3, 4 | 500 | |||
| BC138907.1 | - | 3, 4 | 472 | 8861929 | ||
| BC138909.1 | - | 3, 4 | 472 | 8861931 | ||

Clec16a
Genomic Map