siRNA Details
| Gene Symbol: Ppapdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028922.3 | NP_083198.1 | 1 | 610 | ||
| GenBank mRNA | AK016572.1 | - | 1 | 611 | ||
| AK041581.1 | - | 1 | 593 | |||
| AK149181.1 | - | 1 | 372 | |||
| BC052412.1 | - | 1 | 577 | 5720146 | ||

Ppapdc2
Genomic Map