siRNA Details
Gene Symbol: 4933402D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256158.1 | NP_001243087.1 | 2 | 199 | ||
GenBank mRNA | AK016612.1 | - | 2 | 200 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933402D24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256158.1 | NP_001243087.1 | 2 | 199 | ||
GenBank mRNA | AK016612.1 | - | 2 | 200 |