siRNA Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163262.1 | NP_001156734.1 | 1, 2 | 388 | ||
| GenBank mRNA | AK085367.1 | - | 1, 2 | 291 | ||
| BC158118.1 | - | 1, 2 | 265 | 9088191 | ||
| BY243950.1 | - | 1, 2 | 257 | |||
| BY792820.1 | - | 1, 2 | 269 | |||

Cmip
Genomic Map