siRNA Details
| Gene Symbol: Cfap53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028948.2 | NP_083224.2 | 5 | 950 | ||
| GenBank mRNA | AK016821.1 | - | 6 | 1161 | ||
| BC099500.1 | - | 5 | 868 | 30932292 | ||
| BC125572.1 | - | 5 | 840 | 40130087 | ||
| BC125574.1 | - | 5 | 840 | 40130089 | ||

Cfap53
Genomic Map