siRNA Details
| Gene Symbol: Nsun6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165941.1 | NP_001159413.1 | 8 | 1104 | ||
| NM_001165943.1 | NP_001159415.1 | 7 | 808 | |||
| NM_028950.4 | NP_083226.3 | 7 | 991 | |||
| GenBank mRNA | AK016629.1 | - | 7 | 809 | ||
| AK016811.1 | - | 8 | 1117 | |||
| AK077778.1 | - | 7 | 875 | |||
| AK132428.1 | - | 7 | 958 | |||
| BC025622.1 | - | 4 | 334 | |||
| BC053751.1 | - | 7 | 959 | 6335896 | ||

Nsun6
Genomic Map