siRNA Details
| Gene Symbol: Ifitm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270718.1 | NP_001257647.1 | 2 | 272 | ||
| GenBank mRNA | AK017110.1 | - | 1 | 830 | ||
| AK076846.1 | - | 2 | 272 | |||
| BC115786.1 | - | 1 | 126 | 40105118 | ||
| BC115814.1 | - | 1 | 126 | 40105119 | ||

Ifitm7
Genomic Map