siRNA Details
| Gene Symbol: Sost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024449.6 | NP_077769.4 | 1 | 130 | ||
| GenBank mRNA | AF326740.1 | - | 1 | 88 | ||
| AK017295.1 | - | 1 | 131 | |||
| AK041382.1 | - | 1 | 131 | |||
| BC137832.1 | - | 1 | 120 | 8860851 | ||
| BC137833.1 | - | 1 | 120 | 8860852 | ||

Sost
Genomic Map