siRNA Details
| Gene Symbol: Fam53a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178390.3 | NP_848477.2 | 5 | 1271 | ||
| GenBank mRNA | AF396665.1 | - | 3 | 693 | ||
| AK010537.1 | - | 5 | 1207 | |||
| AK017321.1 | - | 4 | 1616 | |||
| AK155077.1 | - | 6 | 1384 | |||
| AK163326.1 | - | 5 | 1243 | |||
| AK220276.1 | - | 4 | 907 | |||
| BC016596.1 | - | 3 | 696 | 3257498 | ||
| BC028656.1 | - | 5 | 991 | 1244982 | ||

Fam53a
Genomic Map