siRNA Details
| Gene Symbol: Gsdmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028992.1 | NP_083268.1 | 9 | 969 | ||
| GenBank mRNA | AK018537.1 | - | 9 | 970 | ||
| AK163061.1 | - | 9 | 970 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdmc4| Gene Symbol: Gsdmc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028992.1 | NP_083268.1 | 9 | 969 | ||
| GenBank mRNA | AK018537.1 | - | 9 | 970 | ||
| AK163061.1 | - | 9 | 970 | |||