siRNA Details
| Gene Symbol: Ttc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183106.2 | NP_898929.2 | 15 | 1951 | ||
| GenBank mRNA | AK038364.1 | - | 2 | 207 | ||
| BC061503.1 | - | 5 | 524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttc17| Gene Symbol: Ttc17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183106.2 | NP_898929.2 | 15 | 1951 | ||
| GenBank mRNA | AK038364.1 | - | 2 | 207 | ||
| BC061503.1 | - | 5 | 524 | |||