siRNA Details
Gene Symbol: Zkscan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029869.1 | NP_084145.1 | 4 | 1503 | ||
NM_133906.4 | NP_598667.2 | 6 | 1728 | |||
GenBank mRNA | AK028932.1 | - | 6 | 1659 | ||
AK031759.1 | - | 6 | 1707 | |||
AK036807.1 | - | 6 | 1711 | |||
AK054323.1 | - | 6 | 1699 | |||
AK131148.1 | - | 3 | 1648 | |||
AK135999.1 | - | 6 | 1678 | |||
AK154831.1 | - | 6 | 1708 | |||
BC052441.1 | - | 4 | 1467 | 6406518 |
