siRNA Details
| Gene Symbol: Lvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029008.1 | NP_083284.1 | 2 | 872 | ||
| GenBank mRNA | AK014652.1 | - | 2 | 873 | ||
| BC113158.1 | - | 2 | 863 | 40058444 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lvrn| Gene Symbol: Lvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029008.1 | NP_083284.1 | 2 | 872 | ||
| GenBank mRNA | AK014652.1 | - | 2 | 873 | ||
| BC113158.1 | - | 2 | 863 | 40058444 | ||