siRNA Details
| Gene Symbol: Sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029012.2 | NP_083288.2 | 8 | 803 | ||
| GenBank mRNA | AK014709.1 | - | 8 | 1141 | ||
| BC023131.1 | - | 4 | 418 | |||
| BC086646.1 | - | 8 | 1168 | 6817386 | ||
| CD806412.1 | - | 4 | 420 | |||

Sppl3
Genomic Map