siRNA Details
| Gene Symbol: Pomk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029037.4 | NP_083313.1 | 5 | 928 | ||
| GenBank mRNA | AK015374.1 | - | 5 | 921 | ||
| AK032677.1 | - | 5 | 929 | |||
| AK036348.1 | - | 5 | 935 | |||
| AK043620.1 | - | 5 | 929 | |||
| AK138952.1 | - | 5 | 937 | |||
| AK160873.1 | - | 5 | 934 | |||
| AK170994.1 | - | 5 | 929 | |||
| AK202670.1 | - | 1 | 96 | |||
| BC027296.1 | - | 4 | 908 | 4013355 | ||

Pomk
Genomic Map