siRNA Details
| Gene Symbol: Wdr35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159527.1 | NP_001152999.1 | 19 | 2362 | ||
| NM_172470.3 | NP_766058.3 | 20 | 2395 | |||
| GenBank mRNA | AK029896.1 | - | 19 | 2317 | ||
| AK083971.1 | - | 19 | 2307 | |||
| AK173151.1 | - | 20 | 2337 | |||
| BC056925.1 | - | 20 | 2504 | 5704780 | ||

Wdr35
Genomic Map