siRNA Details
| Gene Symbol: Lrrc74b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145435.1 | NP_001138907.1 | 7 | 921 | ||
| NM_029053.3 | NP_083329.2 | 7 | 921 | |||
| GenBank mRNA | AK015438.1 | - | 4 | 1395 | ||
| AK039092.1 | - | 7 | 899 | |||
| AK132588.1 | - | 7 | 919 | |||
| AK143245.1 | - | 7 | 894 | |||
| AK161226.1 | - | 7 | 919 | |||
| BC115678.1 | - | 7 | 898 | 40104166 | ||
| BC115679.1 | - | 7 | 898 | 40104167 | ||

Lrrc74b
Genomic Map