siRNA Details
Gene Symbol: Tbc1d30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029057.1 | NP_083333.1 | 5 | 944 | ||
GenBank mRNA | AK044521.1 | - | 5 | 751 | ||
AK049989.1 | - | 5 | 777 | |||
AK173079.1 | - | 5 | 707 | |||
BC072607.1 | - | 5 | 771 | |||
BC079587.1 | - | 2 | 255 | |||
BC138285.1 | - | 5 | 1151 | 8861304 | ||
BC138286.1 | - | 5 | 1151 | 8861305 |
