siRNA Details
| Gene Symbol: Tbc1d30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029057.1 | NP_083333.1 | 9 | 1509 | ||
| GenBank mRNA | AK015700.1 | - | 3 | 242 | ||
| BC059081.1 | - | 1 | 17 | |||
| BC072607.1 | - | 9 | 1335 | |||
| BC079587.1 | - | 6 | 820 | |||
| BC138285.1 | - | 9 | 1716 | 8861304 | ||
| BC138286.1 | - | 9 | 1716 | 8861305 | ||

Tbc1d30
Genomic Map