siRNA Details
| Gene Symbol: 4930519P11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015848.1 | - | 1 | 369 | ||
| AK053350.1 | - | 1 | 324 | |||
| AK145231.1 | - | 320 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930519P11Rik| Gene Symbol: 4930519P11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015848.1 | - | 1 | 369 | ||
| AK053350.1 | - | 1 | 324 | |||
| AK145231.1 | - | 320 | ||||