siRNA Details
| Gene Symbol: Ddit4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029083.2 | NP_083359.1 | 3 | 776 | ||
| GenBank mRNA | AK017926.1 | - | 3 | 776 | ||
| AK081046.1 | - | 3 | 840 | |||
| AY260552.1 | - | 2 | 699 | |||
| BC131992.1 | - | 3 | 775 | 40130269 | ||
| BC132645.1 | - | 3 | 775 | 40130922 | ||
| BC145370.1 | - | 3 | 775 | 9053928 | ||

Ddit4
Genomic Map