siRNA Details
| Gene Symbol: Sepn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029100.2 | NP_083376.2 | 2 | 290 | ||
| GenBank mRNA | BY742289.1 | - | 2 | 290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sepn1| Gene Symbol: Sepn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029100.2 | NP_083376.2 | 2 | 290 | ||
| GenBank mRNA | BY742289.1 | - | 2 | 290 | ||