siRNA Details
| Gene Symbol: Manf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029103.3 | NP_083379.2 | 3 | 298 | ||
| GenBank mRNA | AB009291.1 | - | 3 | 272 | ||
| AK014338.1 | - | 3 | 276 | |||
| AK131997.1 | - | 3 | 299 | |||
| AK165646.1 | - | 3 | 274 | |||
| AK167580.1 | - | 3 | 298 | |||
| AK176996.1 | - | 3 | 192 | |||
| AK186239.1 | - | 3 | 192 | |||
| BC022761.1 | - | 3 | 304 | |||
| BC038901.1 | - | 3 | 286 | |||
| BC048616.1 | - | 2 | 361 | 6742574 | ||

Manf
Genomic Map